Gene Transcripts

Gene Symbol
Mgrn1
Entrez Gene ID
17237
Gene Name
mahogunin, ring finger 1
Gene Aliases
2610042J20Rik, mKIAA0544, md, nc
Chromosome Location
Chr. 16: 4886100 - 4938296 on Build GRCm38
UniGene ID
Mm.291326
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001252437.1 NP_001239366.1 9 1073
NM_029657.4 NP_083933.1 9 1070
GenBank mRNA AK011747.2 - 9 863
AK088533.1 - 9 853
AK129161.1 - 6 481
AK150176.1 - 10 906
AK153407.1 - 9 894
AK186778.1 - 1 52
BC046830.1 - 9 919 6811606

View Full Product Details