Gene Transcripts

Gene Symbol
Nlrp1a
Entrez Gene ID
195046
Gene Name
NLR family, pyrin domain containing 1A
Gene Aliases
CARD7, DEFCAP, Gm14, Gm15, NAC, Nalp1, Nalp1a, Nlrp1, PP1044
Chromosome Location
Chr. 11: 71091197 - 71144704 on Build GRCm38
UniGene ID
Mm.240227
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004142.2 NP_001004142.2 2, 3 373
GenBank mRNA AK157034.1 - 2, 3 374
AY355339.1 - 1, 2 155
DQ117601.1 - 1, 2 314
KC539856.1 - 1, 2 155
KC539857.1 - 1, 2 155
KC539858.1 - 1, 2 155
KC539859.1 - 1, 2 155
KC539860.1 - 1, 2 155
KC539861.1 - 1, 2 155

View Full Product Details