Gene Transcripts

Gene Symbol
PKDREJ
Entrez Gene ID
10343
Gene Name
polycystin (PKD) family receptor for egg jelly
Gene Aliases
Chromosome Location
Chr. 22: 46255663 - 46263322 on Build GRCh38
UniGene ID
Hs.241383
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006071.1 NP_006062.1 1 2439
GenBank mRNA AF116458.1 - 1 2439

View Full Product Details