Gene Transcripts

Gene Symbol
Slc45a2
Entrez Gene ID
22293
Gene Name
solute carrier family 45, member 2
Gene Aliases
Aim-1, Aim1, Dbr, Matp, Oca4, blanc-sale, bls, uw
Chromosome Location
Chr. 15: 10995693 - 11029233 on Build GRCm38
UniGene ID
Mm.138278
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053077.3 NP_444307.1 1 199
GenBank mRNA AF360357.1 - 1 107
AK029155.1 - 1 200
AK089932.1 - 1 170
AK148207.1 - 196
AK201758.1 - 1 157
AY034377.1 - 1 120
BC125376.1 - 1 186 40129891
BC132429.1 - 1 186 40130706

View Full Product Details