Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_006098.4 | NP_006089.1 | 2 | 255 | |
GenBank mRNA | AK095666.1 | - | 2 | 253 | |
AK222488.1 | - | 2 | 255 | ||
AK301029.1 | - | 2 | 91 | ||
AK301468.1 | - | 2 | 251 | ||
AK303316.1 | - | 2 | 263 | ||
AK310916.1 | - | 2 | 265 | ||
AY336089.1 | - | 2 | 152 | ||
BC000214.1 | - | 2 | 273 | 2959178 | |
BC000366.2 | - | 2 | 243 | 2819249 | |
BC010119.2 | - | 2 | 230 | 3835636 | |
BC014256.1 | - | 2 | 232 | 4765908 | |
BC014788.1 | - | 2 | 239 | 3455986 | |
BC017287.2 | - | 2 | 220 | 4892271 | |
BC019093.2 | - | 2 | 233 | 4893346 | |
BC019362.1 | - | 2 | 237 | 2823761 | |
BC021993.1 | - | 2 | 255 | 4083071 | |
BC029996.1 | - | 2 | 82 | ||
BC032006.1 | - | 2 | 234 | 4750184 | |
BC035460.1 | - | 2 | 236 | ||
CR456978.1 | - | 2 | 149 | ||
CR541909.1 | - | 2 | 149 | ||
GU727632.1 | - | 2 | 149 | ||
M24194.1 | - | 2 | 244 |