Gene Transcripts

Gene Symbol
CFDP1
Entrez Gene ID
10428
Gene Name
craniofacial development protein 1
Gene Aliases
BCNT, BUCENTAUR, CENP-29, CP27, SWC5, Yeti, p97
Chromosome Location
Chr. 16: 75293710 - 75433512 on Build GRCh38
UniGene ID
Hs.461361
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006324.2 NP_006315.1 1, 2 208
XM_011522814.1 XP_011521116.1 1, 2 231
XM_011522815.1 XP_011521117.1 1, 2 231
GenBank mRNA AB009285.1 - 1, 2 180
AK226031.1 - 1, 2 225
AK313948.1 - 1, 2 150
AW160432.1 - 1, 2 212
BC000991.2 - 1, 2 204 3449836
BT009819.1 - 1, 2 71
CR542111.1 - 1, 2 71
D85939.1 - 1, 2 180
LC057679.1 - 1, 2 71
LC057682.1 - 1, 2 71
LC057683.1 - 1, 2 71

View Full Product Details