Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_019660.3 | NP_062634.2 | 5 | 459 | |
GenBank mRNA | AB015858.1 | - | 5 | 422 | |
AK037661.1 | - | 469 | |||
AK077633.1 | - | 5 | 478 | ||
AK077762.1 | - | 5 | 463 | ||
AK077995.1 | - | 5 | 463 | ||
AK132198.1 | - | 324 | |||
AK145773.1 | - | 5 | 460 | ||
BC028344.1 | - | 5 | 457 | 3985166 | |
BC048478.1 | - | 5 | 640 | 6774657 |