Gene Transcripts
- Gene Symbol
- Trav12n-1
- Entrez Gene ID
- 100043218
- Gene Name
- T cell receptor alpha variable 12N-1
- Gene Aliases
- ENSMUSG00000079098, Gm11046
- Chromosome Location
- Chr. 14: 53155607 - 53156088 on Build GRCm38
- UniGene ID
- Mm.333502
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
M38680.1 |
- |
2 |
205 |
|
- Gene Symbol
- Trav12-1
- Entrez Gene ID
- 630086
- Gene Name
- T cell receptor alpha variable 12-1
- Gene Aliases
- Gm17011
- Chromosome Location
- Chr. 14: 53538266 - 53538738 on Build GRCm38
- UniGene ID
- Mm.333502
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC038136.1 |
- |
|
312 |
|
- Gene Symbol
- Trav12-2
- Entrez Gene ID
- 100043322
- Gene Name
- T cell receptor alpha variable 12-2
- Gene Aliases
- Chromosome Location
- Chr. 14: 53616397 - 53616914 on Build GRCm38
- UniGene ID
- Mm.304289
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
U90514.1 |
- |
|
170 |
|
- Gene Symbol
- Trav12n-2
- Entrez Gene ID
- 100043270
- Gene Name
- T cell receptor alpha variable 12N-2
- Gene Aliases
- EG667677, Gm16460
- Chromosome Location
- Chr. 14: 53226884 - 53227400 on Build GRCm38
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK077765.1 |
- |
|
343 |
|
View Full Product Details