Gene Transcripts

Gene Symbol
Leng1
Entrez Gene ID
69757
Gene Name
leukocyte receptor cluster (LRC) member 1
Gene Aliases
1500001K17Rik
Chromosome Location
Chr. 7: 3660107 - 3665840 on Build GRCm38
UniGene ID
Mm.389782
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027203.3 NP_081479.1 2 692
GenBank mRNA AK005095.1 - 2 323
AK037634.1 - 2 339
BC023108.1 - 2 692 4021762

View Full Product Details