Gene Transcripts
- Gene Symbol
- Rpl7a
- Entrez Gene ID
- 27176
- Gene Name
- ribosomal protein L7A
- Gene Aliases
- Surf3
- Chromosome Location
- Chr. 2: 26910807 - 26913311 on Build GRCm38
- UniGene ID
- Mm.328601
- Species
- Mus musculus
- Gene Symbol
- Gm5766
- Entrez Gene ID
- 436332
- Gene Name
- ribosomal protein L7a pseudogene
- Gene Aliases
- EG436332
- Chromosome Location
- Chr. 16: 4229229 - 4231205 on Build GRCm38
- UniGene ID
- Mm.478141
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK030194.1 |
- |
1 |
1867 |
|
BC147642.1 |
- |
1 |
1229 |
|
View Full Product Details