Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_006656.5 | NP_006647.3 | 3 | 1248 | |
GenBank mRNA | AB008185.2 | - | 4 | 1163 | |
AK022450.1 | - | 3 | 1242 | ||
AK290442.1 | - | 3 | 1210 | ||
AK303540.1 | - | 2 | 996 | ||
BC136397.1 | - | 4 | 1582 | 9020384 | |
BC144059.1 | - | 4 | 1582 | 9052579 | |
Y18563.1 | - | 3 | 2019 |