Gene Transcripts

Gene Symbol
RNPS1
Entrez Gene ID
10921
Gene Name
RNA binding protein with serine rich domain 1
Gene Aliases
E5.1
Chromosome Location
Chr. 16: 2253116 - 2268412 on Build GRCh38
UniGene ID
Hs.355643
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001286625.1 NP_001273554.1 3 408
NM_001286626.1 NP_001273555.1 2 415
NM_001286627.1 NP_001273556.1 2 415
NM_006711.4 NP_006702.1 3 469
NM_080594.3 NP_542161.1 3 603
NR_104485.1 2 537
XM_005255048.1 XP_005255105.1 3 696
XM_005255049.3 XP_005255106.1 3 2161
XM_017022874.1 XP_016878363.1 3 1062
GenBank mRNA AB445101.1 - 2 128
AF015608.1 - 2 142
AF274003.1 - 2 104
AK289955.1 - 4 363
AK293343.1 - 2 120
AK303100.1 - 2 118
AK316132.1 - 2 405
AL520572.3 - 3 387
AL559988.3 - 2 191
BC001659.2 - 3 293 2959954
BC001838.2 - 3 293 2959954
BC108316.1 - 3 414 6151864
BG716139.1 - 3 311
BI819738.1 - 2 98
BQ059026.1 - 2 271
DA062271.1 - 3 408
DA728900.1 - 3 305
DQ574166.1 - 1 14
HY012482.1 - 3 470
L37368.1 - 3 677

View Full Product Details