Gene Transcripts

Gene Symbol
KATNA1
Entrez Gene ID
11104
Gene Name
katanin catalytic subunit A1
Gene Aliases
Chromosome Location
Chr. 6: 149594873 - 149649018 on Build GRCh38
UniGene ID
Hs.450175
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001204076.1 NP_001191005.1 5, 6 1017
NM_007044.3 NP_008975.1 7, 8 1245
XM_005266804.2 XP_005266861.1 7, 8 1025
XM_017010207.1 XP_016865696.1 8, 9 1335
XM_017010208.1 XP_016865697.1 6, 7 1061
XM_017010209.1 XP_016865698.1 6, 7 903
XM_017010210.1 XP_016865699.1 7, 8 1030
GenBank mRNA AF056022.1 - 6, 7 941
AK292090.1 - 7, 8 1155
BC050428.1 - 5, 6 854 5764501

View Full Product Details