Gene Transcripts

Gene Symbol
HHLA2
Entrez Gene ID
11148
Gene Name
HERV-H LTR-associating 2
Gene Aliases
B7-H5, B7-H7, B7H7, B7y
Chromosome Location
Chr. 3: 108296490 - 108378285 on Build GRCh38
UniGene ID
Hs.225968
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001282556.1 NP_001269485.1 4 419
NM_001282557.1 NP_001269486.1 4 350
NM_001282558.1 NP_001269487.1 5 497
NM_007072.3 NP_009003.1 4 610
XM_005247080.3 XP_005247137.1 5 480
XM_011512362.1 XP_011510664.1 5 480
XM_011512363.1 XP_011510665.1 4 410
XM_011512364.1 XP_011510666.1 4 361
XM_011512367.2 XP_011510669.1 5 480
XM_017005642.1 XP_016861131.1 4 410
GenBank mRNA AF126162.1 - 4 350
AK000692.1 - 3 345
AK315109.1 - 5 442
BC035971.1 - 4 610 4606764
BP293912.1 - 5 497

View Full Product Details