Gene Transcripts

Gene Symbol
Vps36
Entrez Gene ID
290851
Gene Name
vacuolar protein sorting 36 homolog (S. cerevisiae)
Gene Aliases
RGD1309754
Chromosome Location
Chr. 16: 74719642 - 74743921 on Build Rnor_6.0
UniGene ID
Rn.12707
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106092.2 NP_001099562.2 12 953
GenBank mRNA BC160860.1 - 12 953
FQ217181.1 - 12 964

View Full Product Details