Gene Transcripts

Gene Symbol
Josd2
Entrez Gene ID
292876
Gene Name
Josephin domain containing 2
Gene Aliases
RGD1307305
Chromosome Location
Chr. 1: 100473419 - 100477352 on Build Rnor_6.0
UniGene ID
Rn.22861
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106256.2 NP_001099726.2 5 763
XM_006228996.2 XP_006229058.1 6 1045
XM_006228997.2 XP_006229059.1 6 1042
XM_006228998.2 XP_006229060.1 5 984

View Full Product Details