Gene Transcripts

Gene Symbol
Oxnad1
Entrez Gene ID
306270
Gene Name
oxidoreductase NAD-binding domain containing 1
Gene Aliases
RGD1311534
Chromosome Location
Chr. 16: 8207223 - 8236292 on Build Rnor_6.0
UniGene ID
Rn.3785
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107295.1 NP_001100765.1 6, 7 870
XM_006252623.2 XP_006252685.1 6, 7 971
XM_006252624.2 XP_006252686.1 7, 8 1078
XM_006252625.2 XP_006252687.1 7, 8 1064
XM_006252626.2 XP_006252688.1 6, 7 957
XM_008771000.1 XP_008769222.1 7, 8 1159

View Full Product Details