Gene Transcripts

Gene Symbol
Vkorc1
Entrez Gene ID
309004
Gene Name
vitamin K epoxide reductase complex, subunit 1
Gene Aliases
Chromosome Location
Chr. 1: 199338785 - 199341306 on Build Rnor_6.0
UniGene ID
Rn.97942
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_203335.2 NP_976080.1 1 86
GenBank mRNA AY423047.1 - 1 69
BC166413.1 - 82 9100951
CB314647.1 - 1 102
FQ210919.1 - 1 128

View Full Product Details