Gene Transcripts

Gene Symbol
Alg14
Entrez Gene ID
362031
Gene Name
ALG14, UDP-N-acetylglucosaminyltransferase subunit
Gene Aliases
RGD1312003
Chromosome Location
Chr. 2: 224851352 - 224931461 on Build Rnor_6.0
UniGene ID
Rn.136923
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001014176.2 NP_001014198.1 4 710
XM_008761465.1 XP_008759687.1 5 882
GenBank mRNA BC079151.1 - 4 656

View Full Product Details