Gene Transcripts

Gene Symbol
Cers6
Entrez Gene ID
366065
Gene Name
ceramide synthase 6
Gene Aliases
Lass6, RGD1561599
Chromosome Location
Chr. 3: 55094234 - 55338370 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006234308.2 XP_006234370.1 9 1491
XM_345363.7 XP_345364.5 9 1485

View Full Product Details