Gene Transcripts
- Gene Symbol
- MAN1B1
- Entrez Gene ID
- 11253
- Gene Name
- mannosidase alpha class 1B member 1
- Gene Aliases
- ERMAN1, ERManI, MANA-ER, MRT15
- Chromosome Location
- Chr. 9: 137086862 - 137109187 on Build GRCh38
- UniGene ID
- Hs.279881
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_016219.4 |
NP_057303.2
|
7, 8 |
1149 |
|
NR_045720.1 |
|
7, 8 |
1149 |
|
NR_045721.1 |
|
8, 9 |
1280 |
|
XM_006716945.3 |
XP_006717008.1
|
7, 8 |
1214 |
|
XM_017014239.1 |
XP_016869728.1
|
8, 9 |
1126 |
|
GenBank mRNA |
AF145732.1 |
- |
7, 8 |
1066 |
|
AF148509.1 |
- |
7, 8 |
1126 |
|
AK074699.1 |
- |
5, 6 |
724 |
|
AK130143.1 |
- |
2, 3 |
293 |
|
AK298480.1 |
- |
8, 9 |
1126 |
|
AK299043.1 |
- |
7, 8 |
996 |
|
AK309808.1 |
- |
8, 9 |
1126 |
|
AK315797.1 |
- |
7, 8 |
1091 |
|
AY358465.1 |
- |
7, 8 |
1098 |
|
BC002953.1 |
- |
7, 8 |
1082 |
3533651 |
BC006079.1 |
- |
7, 8 |
1091 |
3959196 |
CR749534.1 |
- |
8, 9 |
1122 |
|
View Full Product Details