Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001199492.1 | NP_001186421.1 | 8 | 1171 | |
NM_014456.4 | NP_055271.2 | 8 | 1213 | ||
NM_145341.3 | NP_663314.1 | 9 | 1297 | ||
GenBank mRNA | AK223623.1 | - | 8 | 1087 | |
AK296753.1 | - | 8 | 1038 | ||
AK312540.1 | - | 8 | 1109 | ||
BC026104.2 | - | 8 | 1080 | 4798293 | |
BC031049.1 | - | 8 | 1080 | 5277621 | |
BX537500.1 | - | 6 | 5086 | ||
BX641168.1 | - | 3 | 3531 | ||
BX647834.1 | - | 8 | 1055 | ||
U96628.1 | - | 8 | 1011 |