Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001031804.2 | NP_001026974.1 | 1 | 1093 | |
NM_005360.4 | NP_005351.2 | 1 | 1093 | ||
XM_017023233.1 | XP_016878722.1 | 1 | 777 | ||
XM_017023234.1 | XP_016878723.1 | 1 | 777 | ||
XM_017023235.1 | XP_016878724.1 | 1 | 777 | ||
GenBank mRNA | AF055376.1 | - | 1 | 1077 | |
AF055377.1 | - | 1 | 1077 | ||
AF540388.1 | - | 1201 | |||
BC038438.1 | - | 1 | 1090 | ||
BC081542.1 | - | 1 | 385 | 30347784 | |
BF726289.1 | - | 1 | 376 |