Gene Transcripts

Gene Symbol
PDGFRL
Entrez Gene ID
5157
Gene Name
platelet derived growth factor receptor like
Gene Aliases
PDGRL, PRLTS
Chromosome Location
Chr. 8: 17576433 - 17643133 on Build GRCh38
UniGene ID
Hs.458573
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006207.2 NP_006198.1 3 547
GenBank mRNA AK289450.1 - 3 547
BC010927.1 - 2 118 4278882
CR541952.1 - 2 102
D37965.1 - 2 163

View Full Product Details