Gene Transcripts

Gene Symbol
EMC1
Entrez Gene ID
23065
Gene Name
ER membrane protein complex subunit 1
Gene Aliases
CAVIPMR, KIAA0090
Chromosome Location
Chr. 1: 19215664 - 19251559 on Build GRCh38
UniGene ID
Hs.439200
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271427.1 NP_001258356.1 23 2870
NM_001271428.1 NP_001258357.1 23 2870
NM_001271429.1 NP_001258358.1 22 2807
NM_015047.2 NP_055862.1 23 2873
XM_005245787.1 XP_005245844.1 24 2877
XM_005245788.1 XP_005245845.1 24 2874
GenBank mRNA AK025853.1 - 10 1234
AK075563.1 - 22 2770
AK124379.1 - 2 1658
AK124740.1 - 3 1305
AK291618.1 - 23 2836
BC034589.1 - 23 2866 4831005
BX648627.1 - 23 2852
BX648708.1 - 23 2828
D42044.2 - 23 2817

View Full Product Details