Gene Transcripts

Gene Symbol
LINC00469
Entrez Gene ID
283982
Gene Name
long intergenic non-protein coding RNA 469
Gene Aliases
C17orf54
Chromosome Location
Chr. 17: 73749270 - 73828537 on Build GRCh38
UniGene ID
Hs.464079
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_027146.1 2 151
GenBank mRNA AK097638.1 - 2 151
BC101213.1 - 2 134
BC101214.1 - 2 134
BC101215.1 - 2 134

View Full Product Details