Gene Transcripts
- Gene Symbol
- SPCS1
- Entrez Gene ID
- 28972
- Gene Name
- signal peptidase complex subunit 1
- Gene Aliases
- HSPC033, SPC1, SPC12, YJR010C-A
- Chromosome Location
- Chr. 3: 52705841 - 52708182 on Build GRCh38
- UniGene ID
- Hs.11125
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_014041.3 |
NP_054760.3
|
2, 3 |
506 |
|
GenBank mRNA |
AF092138.1 |
- |
2, 3 |
235 |
|
AK027426.1 |
- |
2, 3 |
235 |
|
AX775783.1 |
- |
2, 3 |
235 |
|
AX775785.1 |
- |
3, 4 |
1020 |
|
AX775787.1 |
- |
2, 3 |
209 |
|
AX775833.1 |
- |
2, 3 |
225 |
|
BC000884.1 |
- |
2, 3 |
209 |
3462307 |
BQ423345.1 |
- |
2, 3 |
242 |
|
L38852.1 |
- |
2, 3 |
225 |
|
View Full Product Details