Gene Transcripts
- Gene Symbol
- SPDYE7P
- Entrez Gene ID
- 441251
- Gene Name
- speedy/RINGO cell cycle regulator family member E7, pseudogene
- Gene Aliases
- Chromosome Location
- Chr. 7: 72862757 - 72869091 on Build GRCh38
- UniGene ID
- Hs.591819
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_003666.2 |
|
3 |
404 |
|
GenBank mRNA |
BC100974.2 |
- |
3 |
404 |
|
View Full Product Details