Gene Transcripts
- Gene Symbol
- MIIP
- Entrez Gene ID
- 60672
- Gene Name
- migration and invasion inhibitory protein
- Gene Aliases
- IIP45
- Chromosome Location
- Chr. 1: 12019242 - 12032049 on Build GRCh38
- UniGene ID
- Hs.8595
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_021933.3 |
NP_068752.2
|
2, 3 |
513 |
|
XM_005263487.3 |
XP_005263544.1
|
2, 3 |
342 |
|
XM_011541895.1 |
XP_011540197.1
|
2, 3 |
344 |
|
XM_011541896.1 |
XP_011540198.1
|
2, 3 |
237 |
|
GenBank mRNA |
AK022500.1 |
- |
2, 3 |
293 |
|
AK024020.1 |
- |
2, 3 |
232 |
|
BC008068.2 |
- |
2, 3 |
222 |
3050071 |
BG536799.1 |
- |
2, 3 |
301 |
|
BI828925.1 |
- |
2, 3 |
270 |
|
FJ618905.1 |
- |
2, 3 |
513 |
|
View Full Product Details