Gene Transcripts

Gene Symbol
LIMCH1
Entrez Gene ID
22998
Gene Name
LIM and calponin homology domains 1
Gene Aliases
LIMCH1A, LMO7B
Chromosome Location
Chr. 4: 41359607 - 41700044 on Build GRCh38
UniGene ID
Hs.335163
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001112717.2 NP_001106188.1 24 3054
NM_001112718.2 NP_001106189.1 24 3051
NM_001112719.2 NP_001106190.1 19 2836
NM_001112720.2 NP_001106191.1 18 2800
NM_001289122.1 NP_001276051.1 24 3090
NM_001289124.1 NP_001276053.1 24 3135
NM_014988.3 NP_055803.2 25 3132
XM_005248057.1 XP_005248114.1 32 4773
XM_005248058.1 XP_005248115.1 32 4770
XM_005248060.1 XP_005248117.1 32 4761
XM_005248061.1 XP_005248118.1 31 4689
XM_005248067.2 XP_005248124.1 30 5635
XM_005248072.1 XP_005248129.1 25 3129
XM_005248074.1 XP_005248131.1 24 3093
XM_005248075.1 XP_005248132.1 23 3015
XM_006713996.1 XP_006714059.1 31 4683
XM_011513642.1 XP_011511944.1 33 4848
XM_011513643.1 XP_011511945.1 33 4845
XM_011513644.1 XP_011511946.1 33 4839
XM_011513645.1 XP_011511947.1 32 4764
XM_011513646.1 XP_011511948.1 32 4761
XM_011513648.1 XP_011511950.1 32 4605
XM_011513649.1 XP_011511951.1 28 4507
XM_011513651.2 XP_011511953.1 31 5727
XM_011513653.1 XP_011511955.1 26 3216
XM_011513654.1 XP_011511956.1 25 3180
XM_011513655.1 XP_011511957.1 20 2839
XM_011513656.1 XP_011511958.1 20 2797
XM_011513657.1 XP_011511959.1 19 2752
XM_011513658.2 XP_011511960.1 23 3276
XM_017007894.1 XP_016863383.1 33 4836
XM_017007895.1 XP_016863384.1 32 4764
XM_017007896.1 XP_016863385.1 31 4677
XM_017007897.1 XP_016863386.1 30 4602
XM_017007898.1 XP_016863387.1 25 3141
XM_017007899.1 XP_016863388.1 25 3138
XM_017007900.1 XP_016863389.1 24 3057
XM_017007901.1 XP_016863390.1 24 3048
XM_017007902.1 XP_016863391.1 23 2970
XM_017007903.1 XP_016863392.1 21 2875
XM_017007904.1 XP_016863393.1 20 2788
XM_017007905.1 XP_016863394.1 19 2713
GenBank mRNA AB029025.2 - 25 3130
AK027231.1 - 11 1056
AK294774.1 - 19 2941
AK295784.1 - 18 2467
AK295836.1 - 19 2577
AK299297.1 - 18 2670
AK299835.1 - 19 2612
AK302674.1 - 11 1302
AK309846.1 - 18 2742
AL831962.2 - 30 4516
BC023546.2 - 21 2904 4110496
BC029735.1 - 9 806
BC095394.1 - 14 1351
BX640692.1 - 17 2428
CR749205.1 - 19 2706
CR933645.1 - 24 3055
CR936601.1 - 24 3061
CR936610.1 - 24 3136
CR936658.1 - 21 2802
CR936664.1 - 24 3052

View Full Product Details