Gene Transcripts
- Gene Symbol
- VASH2
- Entrez Gene ID
- 79805
- Gene Name
- vasohibin 2
- Gene Aliases
- Chromosome Location
- Chr. 1: 212935543 - 212991585 on Build GRCh38
- UniGene ID
- Hs.96885
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001136474.2 |
NP_001129946.1
|
7, 8 |
1106 |
|
NM_001136475.2 |
NP_001129947.1
|
5, 6 |
822 |
|
NM_001301056.1 |
NP_001287985.1
|
6, 7 |
1302 |
|
NM_024749.4 |
NP_079025.2
|
4, 5 |
1170 |
|
XM_011509987.2 |
XP_011508289.1
|
9, 10 |
1586 |
|
XM_011509988.2 |
XP_011508290.1
|
10, 11 |
1660 |
|
XM_011509989.2 |
XP_011508291.1
|
6, 7 |
1280 |
|
XM_011509991.2 |
XP_011508293.2
|
5, 6 |
999 |
|
XM_011509995.1 |
XP_011508297.1
|
4, 5 |
702 |
|
XM_017002351.1 |
XP_016857840.1
|
7, 8 |
1420 |
|
XM_017002352.1 |
XP_016857841.1
|
4, 5 |
853 |
|
GenBank mRNA |
AK022567.1 |
- |
7, 8 |
1032 |
|
AK096605.1 |
- |
7, 8 |
970 |
|
AK290973.1 |
- |
7, 8 |
1065 |
|
AK302675.1 |
- |
5, 6 |
675 |
|
AY834202.1 |
- |
5, 6 |
877 |
|
BC051856.1 |
- |
4, 5 |
1055 |
6067012 |
View Full Product Details