Gene Transcripts
- Gene Symbol
- SH3PXD2A
- Entrez Gene ID
- 9644
- Gene Name
- SH3 and PX domains 2A
- Gene Aliases
- FISH, SH3MD1, TKS5
- Chromosome Location
- Chr. 10: 103594027 - 103855464 on Build GRCh38
- UniGene ID
- Hs.678727
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_014631.2 |
NP_055446.2
|
13, 14 |
1492 |
|
XM_005270294.4 |
XP_005270351.1
|
14, 15 |
1625 |
|
XM_005270295.4 |
XP_005270352.1
|
13, 14 |
1579 |
|
XM_005270297.3 |
XP_005270354.1
|
8, 9 |
2856 |
|
XM_005270298.4 |
XP_005270355.1
|
8, 9 |
1659 |
|
XM_006718079.3 |
XP_006718142.1
|
15, 16 |
1722 |
|
XM_006718080.3 |
XP_006718143.1
|
9, 10 |
1753 |
|
XM_011540385.2 |
XP_011538687.1
|
14, 15 |
1676 |
|
XM_011540386.2 |
XP_011538688.1
|
9, 10 |
2952 |
|
XM_011540387.2 |
XP_011538689.1
|
5, 6 |
4598 |
|
XM_017016969.1 |
XP_016872458.1
|
14, 15 |
1643 |
|
XM_017016970.1 |
XP_016872459.1
|
13, 14 |
1459 |
|
GenBank mRNA |
AB007878.1 |
- |
11, 12 |
1242 |
|
AK056469.1 |
- |
12, 13 |
1215 |
|
AK294454.1 |
- |
8, 9 |
1032 |
|
AK294736.1 |
- |
8, 9 |
1070 |
|
AK295660.1 |
- |
8, 9 |
1136 |
|
AK302015.1 |
- |
11, 12 |
1248 |
|
AK304828.1 |
- |
8, 9 |
1048 |
|
AK315983.1 |
- |
8, 9 |
1075 |
|
View Full Product Details