Gene Transcripts

Gene Symbol
MALRD1
Entrez Gene ID
340895
Gene Name
MAM and LDL receptor class A domain containing 1
Gene Aliases
C10orf112, DIET1, bA265G8.2
Chromosome Location
Chr. 10: 19046931 - 19734478 on Build GRCh38
UniGene ID
Hs.646734
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001142308.2 NP_001135780.2 34 5882
XM_011519453.2 XP_011517755.1 35 6420
XM_011519454.1 XP_011517756.1 35 5746
XM_011519455.2 XP_011517757.1 34 6247
XM_017016182.1 XP_016871671.1 34 5692
XM_017016183.1 XP_016871672.1 35 6421
XM_017016184.1 XP_016871673.1 23 4109
XM_017016185.1 XP_016871674.1 23 4106
XM_017016186.1 XP_016871675.1 16 2663
GenBank mRNA AK297683.1 - 6 765
KC843478.1 - 34 5882

View Full Product Details