Gene Transcripts
- Gene Symbol
- GGA2
- Entrez Gene ID
- 23062
- Gene Name
- golgi associated, gamma adaptin ear containing, ARF binding protein 2
- Gene Aliases
- VEAR
- Chromosome Location
- Chr. 16: 23463542 - 23510494 on Build GRCh38
- UniGene ID
- Hs.460336
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_015044.4 |
NP_055859.1
|
16, 17 |
1819 |
|
XM_017023074.1 |
XP_016878563.1
|
17, 18 |
5508 |
|
XM_017023075.1 |
XP_016878564.1
|
16, 17 |
1822 |
|
GenBank mRNA |
AB029003.1 |
- |
13, 14 |
1449 |
|
AF165531.1 |
- |
16, 17 |
1757 |
|
AF190863.1 |
- |
16, 17 |
1763 |
|
AF233522.1 |
- |
16, 17 |
1762 |
|
AF323754.1 |
- |
17, 18 |
1675 |
|
AK024619.1 |
- |
16, 17 |
1818 |
|
AK093769.1 |
- |
1, 2 |
702 |
|
BC000284.1 |
- |
16, 17 |
1801 |
3356490 |
View Full Product Details