Gene Transcripts

Gene Symbol
SPANXB1
Entrez Gene ID
728695
Gene Name
SPANX family member B1
Gene Aliases
B1, CT11.2, SPANX-B, SPANXB, SPANXB2, SPANXF1, SPANXF2
Chromosome Location
Chr. X: 141002591 - 141003706 on Build GRCh38
UniGene ID
Hs.434105
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032461.3 NP_115850.2 2 411
GenBank mRNA AF098307.1 - 2 411
AJ457796.1 - 2 411
BC034472.1 - 2 409 4824918
BC137533.1 - 2 401 9021533
BC137536.1 - 2 401 9021536

View Full Product Details