Gene Transcripts

Gene Symbol
PIK3IP1
Entrez Gene ID
113791
Gene Name
phosphoinositide-3-kinase interacting protein 1
Gene Aliases
HGFL, hHGFL(S)
Chromosome Location
Chr. 22: 31281593 - 31292534 on Build GRCh38
UniGene ID
Hs.26670
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_052880.4 NP_443112.2 5 718
GenBank mRNA AF528079.1 - 5 663
AF528080.1 - 5 663
AK074688.1 - 5 664
AK093768.1 - 2 1278
AK226089.1 - 5 598
AK300990.1 - 6 576
BC011049.2 - 5 663 4180134
BC041903.1 - 3 1015
BE299406.1 - 3 346
BQ718653.1 - 5 696
CR456340.1 - 5 558
GU135609.1 - 3 342

View Full Product Details