Gene Transcripts

Gene Symbol
SEPT8
Entrez Gene ID
23176
Gene Name
septin 8
Gene Aliases
SEP2
Chromosome Location
Chr. 5: 132750817 - 132778216 on Build GRCh38
UniGene ID
Hs.522057
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001098811.1 NP_001092281.1 4 692
NM_001098812.1 NP_001092282.1 4 692
NM_001098813.1 NP_001092283.1 4 618
NM_001300798.1 NP_001287727.1 4 686
NM_001300799.1 NP_001287728.1 4 686
NM_015146.1 NP_055961.1 4 692
XM_005271927.2 XP_005271984.1 4 694
XM_005271928.2 XP_005271985.1 4 688
XM_005271931.3 XP_005271988.1 5 775
XM_011543288.1 XP_011541590.1 4 714
XM_017009254.1 XP_016864743.1 4 688
XM_017009255.1 XP_016864744.1 4 761
XM_017009256.1 XP_016864745.1 4 755
XM_017009257.1 XP_016864746.1 4 708
XM_017009258.1 XP_016864747.1 4 761
XM_017009259.1 XP_016864748.1 4 755
XM_017009260.1 XP_016864749.1 4 708
XM_017009261.1 XP_016864750.1 4 755
GenBank mRNA AF179995.1 - 4 692
AF440761.1 - 4 692
AF440762.1 - 4 692
AF440763.1 - 4 527
AK057797.1 - 4 550
BC001329.1 - 1 91
BC140759.1 - 4 508 9021627
BC171779.1 - 4 508 9054433
D86957.1 - 4 692

View Full Product Details