Gene Transcripts

Gene Symbol
Iqcf6
Entrez Gene ID
100041096
Gene Name
IQ motif containing F6
Gene Aliases
Chromosome Location
Chr. 9: 106626582 - 106627675 on Build GRCm38
UniGene ID
Mm.477435
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001101628.1 NP_001095098.1 2 217

View Full Product Details