Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_029751.4 | NP_084027.1 | 2 | 151 | |
GenBank mRNA | AK010983.1 | - | 2 | 122 | |
AK146543.1 | - | 2 | 121 | ||
AK150820.1 | - | 2 | 120 | ||
AK152611.1 | - | 2 | 120 | ||
AK161942.1 | - | 2 | 121 | ||
AK162047.1 | - | 2 | 122 | ||
BC024609.1 | - | 2 | 108 | 4221597 | |
BC037146.1 | - | 2 | 163 | 5011713 | |
CJ042325.1 | - | 2 | 152 |