Gene Transcripts

Gene Symbol
Znhit1
Entrez Gene ID
70103
Gene Name
zinc finger, HIT domain containing 1
Gene Aliases
2700001K05Rik, ZNFN4A1
Chromosome Location
Chr. 5: 136982194 - 136987882 on Build GRCm38
UniGene ID
Mm.273147
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027318.3 NP_081594.1 3 590
GenBank mRNA AK012199.1 - 3 230
AK080799.1 - 3 591
BC026751.1 - 3 236 3153902

View Full Product Details