Gene Transcripts

Gene Symbol
Trim7
Entrez Gene ID
94089
Gene Name
tripartite motif-containing 7
Gene Aliases
AI790312
Chromosome Location
Chr. 11: 48826138 - 48850644 on Build GRCm38
UniGene ID
Mm.390335
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053166.2 NP_444396.2 3 413
GenBank mRNA AF396656.1 - 3 428

View Full Product Details