Gene Transcripts

Gene Symbol
Mettl7a2
Entrez Gene ID
393082
Gene Name
methyltransferase like 7A2
Gene Aliases
Gm921, UbiE2, Ubie
Chromosome Location
Chr. 15: 100353200 - 100361819 on Build GRCm38
UniGene ID
Mm.268451
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_199477.2 NP_955771.2 2 1572
GenBank mRNA AB253944.1 - 2 1563
AK143923.1 - 2 1573
BC090971.1 - 2 1572 30314995

View Full Product Details