Gene Transcripts

Gene Symbol
Sprr2b
Entrez Gene ID
20756
Gene Name
small proline-rich protein 2B
Gene Aliases
Chromosome Location
Chr. 3: 92316705 - 92318085 on Build GRCm38
UniGene ID
Mm.445310
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011469.3 NP_035599.2 385
GenBank mRNA AY158987.1 - 385
AY298855.1 - 2 459
BC132518.1 - 371 40130795

View Full Product Details