Gene Transcripts

Gene Symbol
Vmn2r97
Entrez Gene ID
627367
Gene Name
vomeronasal 2, receptor 97
Gene Aliases
EG627367, Vn2r97
Chromosome Location
Chr. 17: 18914322 - 18948071 on Build GRCm38
UniGene ID
Mm.384131
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001104549.1 NP_001098019.1 3 1318

View Full Product Details