Gene Transcripts

Gene Symbol
Rhox3a
Entrez Gene ID
382209
Gene Name
reproductive homeobox 3A
Gene Aliases
AY147207, Gm1138, Prx, Rhox3, Rhox3.1
Chromosome Location
Chr. X: 37249919 - 37258978 on Build GRCm38
UniGene ID
Mm.357753
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_194063.3 NP_918952.2 3 263
GenBank mRNA DQ058640.1 - 3 263

Gene Symbol
Rhox3c
Entrez Gene ID
100135654
Gene Name
reproductive homeobox 3C
Gene Aliases
EG665203, Rhox3.3, Rhox3b
Chromosome Location
Chr. X: 37469868 - 37473961 on Build GRCm38
UniGene ID
Mm.359489
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001102457.4 NP_001095927.3 1 43

Gene Symbol
Rhox3e
Entrez Gene ID
100135657
Gene Name
reproductive homeobox 3E
Gene Aliases
ENSMUSG00000071772, OTTMUSG00000017148, Rhox3.5, Rhox3d
Chromosome Location
Chr. X: 37546975 - 37550897 on Build GRCm38
UniGene ID
Mm.379498
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001184969.1 NP_001171898.1 43

Gene Symbol
Rhox3f
Entrez Gene ID
621852
Gene Name
reproductive homeobox 3F
Gene Aliases
OTTMUSG00000017155, Rhox3.6, Rhox3e
Chromosome Location
Chr. X: 37581352 - 37585496 on Build GRCm38
UniGene ID
Mm.490237
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001040089.3 NP_001035178.3 1 43

Gene Symbol
Rhox3h
Entrez Gene ID
434758
Gene Name
reproductive homeobox 3H
Gene Aliases
EG434758, Rhox3.8, Rhox3g
Chromosome Location
Chr. X: 37657688 - 37668764 on Build GRCm38
UniGene ID
Mm.357753
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001114157.1 NP_001107629.1 3 263

View Full Product Details