Gene Transcripts

Gene Symbol
Ssxb9
Entrez Gene ID
387131
Gene Name
synovial sarcoma, X member B, breakpoint 9
Gene Aliases
Chromosome Location
Chr. X: 8366978 - 8375388 on Build GRCm38
UniGene ID
Mm.379565
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_199066.2 NP_951021.2 6 555
GenBank mRNA AY244800.1 - 555

View Full Product Details