Gene Transcripts
- Gene Symbol
- Mup10
- Entrez Gene ID
- 100039008
- Gene Name
- major urinary protein 10
- Gene Aliases
- 2610016E04Rik, Mup8
- Chromosome Location
- Chr. 4: 60578260 - 60582155 on Build GRCm38
- UniGene ID
- Mm.335875
- Species
- Mus musculus
- Gene Symbol
- LOC100048884
- Entrez Gene ID
- 100048884
- Gene Name
- novel member of the major urinary protein (Mup) gene family
- Gene Aliases
- Mup6
- Chromosome Location
- Chr. 4: 61670177 - 61674098 on Build GRCm38
- UniGene ID
- Mm.335875
- Species
- Mus musculus
- Gene Symbol
- Mup1
- Entrez Gene ID
- 17840
- Gene Name
- major urinary protein 1
- Gene Aliases
- Ltn-1, Lvtn-1, Mup-1, Mup-a, Mup10, Mup7, Up-1
- Chromosome Location
- Chr. 4: 60498012 - 60501960 on Build GRCm38
- UniGene ID
- Mm.335875
- Species
- Mus musculus
- Gene Symbol
- Mup11
- Entrez Gene ID
- 100039028
- Gene Name
- major urinary protein 11
- Gene Aliases
- Gm12549, OTTMUSG00000007431
- Chromosome Location
- Chr. 4: 60658466 - 60662395 on Build GRCm38
- UniGene ID
- Mm.482557
- Species
- Mus musculus
- Gene Symbol
- Mup12
- Entrez Gene ID
- 100039054
- Gene Name
- major urinary protein 12
- Gene Aliases
- Gm2024
- Chromosome Location
- Chr. 4: 60737381 - 60741281 on Build GRCm38
- UniGene ID
- Mm.482236
- Species
- Mus musculus
- Gene Symbol
- Mup14
- Entrez Gene ID
- 100039116
- Gene Name
- major urinary protein 14
- Gene Aliases
- Gm13514, Mup12, OTTMUSG00000012493
- Chromosome Location
- Chr. 4: 61300037 - 61303998 on Build GRCm38
- UniGene ID
- Mm.458441
- Species
- Mus musculus
- Gene Symbol
- Mup15
- Entrez Gene ID
- 100039150
- Gene Name
- major urinary protein 15
- Gene Aliases
- Gm2068, Mup13
- Chromosome Location
- Chr. 4: 61435787 - 61439714 on Build GRCm38
- UniGene ID
- Mm.237772
- Species
- Mus musculus
- Gene Symbol
- Mup16
- Entrez Gene ID
- 100039177
- Gene Name
- major urinary protein 16
- Gene Aliases
- Chromosome Location
- Chr. 4: 61515592 - 61519503 on Build GRCm38
- UniGene ID
- Mm.422695
- Species
- Mus musculus
- Gene Symbol
- Mup19
- Entrez Gene ID
- 100189605
- Gene Name
- major urinary protein 19
- Gene Aliases
- 100039247, Gm12552, Mup11, Mup14, Mup17, Mup8, OTTMUSG00000007472
- Chromosome Location
- Chr. 4: 61778324 - 61782236 on Build GRCm38
- UniGene ID
- Mm.457980
- Species
- Mus musculus
- Gene Symbol
- Mup2
- Entrez Gene ID
- 17841
- Gene Name
- major urinary protein 2
- Gene Aliases
- AA589603, Mup-2, Mup4
- Chromosome Location
- Chr. 4: 60135913 - 60139872 on Build GRCm38
- UniGene ID
- Mm.335875
- Species
- Mus musculus
- Gene Symbol
- Mup8
- Entrez Gene ID
- 100041687
- Gene Name
- major urinary protein 8
- Gene Aliases
- Gm12809, Mup5, OTTMUSG00000008509
- Chromosome Location
- Chr. 4: 60218620 - 60222599 on Build GRCm38
- UniGene ID
- Mm.457982
- Species
- Mus musculus
- Gene Symbol
- Mup9
- Entrez Gene ID
- 100038948
- Gene Name
- major urinary protein 9
- Gene Aliases
- Gm14076, Mup2, Mup6, OTTMUSG00000015595
- Chromosome Location
- Chr. 4: 60418046 - 60421961 on Build GRCm38
- UniGene ID
- Mm.482231
- Species
- Mus musculus
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BC094452.1 |
- |
|
194 |
|
Chr. - - [-] |
View Full Product Details