Gene Transcripts

Gene Symbol
Pitx3
Entrez Gene ID
29609
Gene Name
paired-like homeodomain 3
Gene Aliases
Chromosome Location
Chr. 1: 265886766 - 265899947 on Build Rnor_6.0
UniGene ID
Rn.22092
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019247.1 NP_062120.1 3 310
XM_006231478.2 XP_006231540.1 3 489
GenBank mRNA AJ011005.2 - 3 310

View Full Product Details