Gene Transcripts

Gene Symbol
SATB2
Entrez Gene ID
23314
Gene Name
SATB homeobox 2
Gene Aliases
GLSS
Chromosome Location
Chr. 2: 199269500 - 199471266 on Build GRCh38
UniGene ID
Hs.516617
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001172509.1 NP_001165980.1 7 1656
NM_001172517.1 NP_001165988.1 8 1252
NM_015265.3 NP_056080.1 8 1232
XM_005246396.2 XP_005246453.1 6 887
XM_006712372.2 XP_006712435.1 7 1002
XM_011510840.2 XP_011509142.1 8 1226
XM_017003656.1 XP_016859145.1 7 887
GenBank mRNA AB028957.1 - 7 925
AB209376.1 - 8 1252
AK025127.1 - 1 142
AK056638.1 - 7 1657
AK291463.1 - 8 1238
BC098136.1 - 6 890 40007830
BC099723.1 - 6 890 40007832
BC103492.1 - 3 536 40007829
BC103500.1 - 6 890 40007831

View Full Product Details