Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001292038.1 | NP_001278967.1 | 5 | 668 | |
NM_015274.2 | NP_056089.1 | 5 | 668 | ||
GenBank mRNA | AB023152.2 | - | 5 | 627 | |
AK301473.1 | - | 5 | 653 | ||
AL832232.1 | - | 5 | 638 | ||
AL833071.2 | - | 5 | 641 | ||
BC033307.1 | - | 5 | 637 | 4932398 | |
BC094773.1 | - | 5 | 668 | 30531368 |